Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((2-Acetamido-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl acetate 97%

Supplier: AMBEED, INC
Warning

A620616-100G A620616-1G A620616-25G A620616-5G
76727-064EA 44 USD InStock
76727-064 76727-066 76727-068 76727-070
2-((2-Acetamido-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl acetate 97%
2-((2-Acetamido-6-oxo-1H-purin-9(6H)-yl)methoxy)ethyl acetate
Precaución: for Research Use Only


Formula: C₁₂H₁₅N₅O₅
MDL Number: MFCD00267706
CAS Number: 75128-73-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty