Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Phthaloyl-L-glutamic anhydride 95%

Supplier: AMBEED, INC
Warning

A629646-100G A629646-25G A629646-5G
76727-140EA 109 USD InStock
76727-140 76727-142 76727-144
N-Phthaloyl-L-glutamic anhydride 95%
N-Phthaloyl-L-glutamic anhydride
Precaución: for Research Use Only


Formula: C₁₃H₉NO₅
MDL Number: MFCD00067324
CAS Number: 25830-77-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty