Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium propionate 99+%

Supplier: AMBEED, INC
Warning

A631578-100G A631578-500G
76727-154EA 9 USD InStock
76727-154 76727-156 76727-158 76727-160
Sodium propionate 99+%
Sodium propionate
Precaución: for Research Use Only


Formula: C₃H₅NaO₂
MW: 96.06 g/mol
Melting Pt: 288.76
Flash Pt: 157
MDL Number: MFCD00002759
CAS Number: 137-40-6
Merck Index: 12,08816
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty