Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dibenzyl-2-aminoethanol 98%

Supplier: AMBEED, INC

A639618-100G A639618-25G A639618-500G A639618-5G
76727-222EA 50 USD InStock
76727-222 76727-224 76727-226 76727-228
N,N-Dibenzyl-2-aminoethanol 98%
N,N-Dibenzyl-2-aminoethanol
Precaución: for Research Use Only


Formula: C₁₆H₁₉NO
MDL Number: MFCD00020574
CAS Number: 101-06-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty