Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-4'-fluorobiphenyl 95%

Supplier: AMBEED, INC
Danger

A640068-100MG A640068-1G A640068-250MG A640068-5G
76727-234EA 8 USD InStock
76727-234 76727-236 76727-238 76727-240
4-Bromo-4'-fluorobiphenyl 95%
4-Bromo-4'-fluorobiphenyl
Precaución: for Research Use Only


Formula: C₁₂H₈BrF
MW: 251.1
MDL Number: MFCD00017954
CAS Number: 398-21-0
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty