Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,2-Dicyclohexylethyl)piperidine maleate 98%

Supplier: AMBEED, INC
Warning

A645812-10MG A645812-1MG A645812-5MG
76727-296EA 119 USD InStock
76727-296 76727-298 76727-300
2-(2,2-Dicyclohexylethyl)piperidine maleate 98%
2-(2,2-Dicyclohexylethyl)piperidine maleate
Precaución: for Research Use Only


Formula: C₂₃H₃₉NO₄
MDL Number: MFCD00057329
CAS Number: 6724-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty