Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o-Phenylenediacetonitrile 98%

Supplier: AMBEED, INC
Danger

A646792-1G A646792-25G
76727-316EA 6 USD InStock
76727-316 76727-318
o-Phenylenediacetonitrile 98%
o-Phenylenediacetonitrile
Precaución: for Research Use Only


Formula: C₁₀H₈N₂
MW: 156.19 g/mol
MDL Number: MFCD00001905
CAS Number: 613-73-0
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty