Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-7-methoxybenzofuran-2-carboxylat 98%

Supplier: AMBEED, INC
Warning

A650514-1G A650514-250MG A650514-25G A650514-5G
76727-336EA 15 USD InStock
76727-336 76727-338 76727-340 76727-342
Ethyl-7-methoxybenzofuran-2-carboxylat 98%
Ethyl 7-methoxybenzofuran-2-carboxylat
Precaución: for Research Use Only


Formula: C₁₂H₁₂O₄
MDL Number: MFCD00060510
CAS Number: 50551-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty