Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Cbz-3-piperidineacetic Acid 97%

Supplier: AMBEED, INC
Warning

A656472-10G A656472-1G A656472-250MG
76727-376EA 112 USD InStock
76727-376 76727-378 76727-380
N-Cbz-3-piperidineacetic Acid 97%
N-Cbz-3-piperidineacetic Acid
Precaución: for Research Use Only


Formula: C₁₅H₁₉NO₄
MDL Number: MFCD02179002
CAS Number: 86827-10-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty