Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Octylamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A656849-1G A656849-250MG A656849-5G
76727-384EA 33 USD InStock
76727-384 76727-386 76727-388
n-Octylamine hydrochloride 98%
n-Octylamine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₉N·HCl
MW: 165.71 g/mol
MDL Number: MFCD00042019
CAS Number: 142-95-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty