Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl piperazine-1-carboxylate 98%

Supplier: AMBEED, INC
Danger

A679667-1G A679667-25G A679667-5G
76727-568EA 59.42 USD InStock
76727-568 76727-570 76727-572 76727-574
Methyl piperazine-1-carboxylate 98%
Methyl piperazine-1-carboxylate
Precaución: for Research Use Only


Formula: C₆H₁₂N₂O₂
MW: 144.172 g/mol
MDL Number: MFCD06411658
CAS Number: 50606-31-0
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty