Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentyltriethoxysilane 98%

Supplier: AMBEED, INC
Warning

A703626-1G A703626-25G A703626-5G
76727-708EA 10 USD InStock
76727-708 76727-710 76727-712
Pentyltriethoxysilane 98%
Pentyltriethoxysilane
Precaución: for Research Use Only


Formula: C₁₁H₂₆O₃Si
MDL Number: MFCD00026757
CAS Number: 2761-24-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty