Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Chloromethyl)benzoyl chloride 95%

Supplier: AMBEED, INC
Danger

A711316-100MG A711316-250MG A711316-5G
76727-780EA 7 USD InStock
76727-780 76727-782 77669-914
2-(Chloromethyl)benzoyl chloride 95%
2-(Chloromethyl)benzoyl chloride
Precaución: For research use only.


Formula: C₈H₆Cl₂O
MDL Number: MFCD00152348
CAS Number: 42908-86-1
UN: 2927
ADR: 6.1,8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty