Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Pentacosane 98%

Supplier: AMBEED, INC
Danger

A715092-100MG A715092-1G A715092-250MG A715092-5G
76727-806EA 32 USD InStock
76727-806 76727-808 76727-810 76727-812
n-Pentacosane 98%
n-Pentacosane
Precaución: for Research Use Only


Formula: H₃C(CH₂)₂₃CH₃
MW: 352.69 g/mol
Boiling Pt: 402
Melting Pt: 54
MDL Number: MFCD00009353
CAS Number: 629-99-2
UN: 3082
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty