Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetic 2-(2,5-dioxopyrrolidin-1-yl)propanoic thioanhydride 98%

Supplier: AMBEED, INC
Warning

A715742-100MG A715742-1G A715742-250MG
76727-822EA 13 USD InStock
76727-822 76727-824 76727-826
Acetic 2-(2,5-dioxopyrrolidin-1-yl)propanoic thioanhydride 98%
Acetic 2-(2,5-dioxopyrrolidin-1-yl)propanoic thioanhydride
Precaución: for Research Use Only


Formula: C₉H₁₁NO₄S
MDL Number: MFCD11519182
CAS Number: 1093759-04-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty