Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methylethylenediamine 97%

Supplier: AMBEED, INC
Danger

A716894-1G A716894-5G
76727-842EA 18 USD InStock
76727-842 76727-844
N-Methylethylenediamine 97%
N-Methylethylenediamine
Precaución: for Research Use Only


Formula: C₃H₁₀N₂
MDL Number: MFCD00008165
CAS Number: 109-81-9
UN: 2734
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty