Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-4-(ethylthio)phenyltrifluoroborate 98%

Supplier: AMBEED, INC
Warning

A718804-1G A718804-25G A718804-5G
76727-860EA 13 USD InStock
76727-860 76727-862 76727-864
Potassium-4-(ethylthio)phenyltrifluoroborate 98%
Potassium 4-(ethylthio)phenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₈H₉BF₃KS
MW: 244.12 g/mol
MDL Number: MFCD04115771
CAS Number: 850623-75-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty