Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Hydroxy-1,8-naphthalimide 97%

Supplier: AMBEED, INC
Warning

A723814-1G A723814-5G
76727-908EA 7 USD InStock
76727-908 76727-910
N-Hydroxy-1,8-naphthalimide 97%
N-Hydroxy-1,8-naphthalimide
Precaución: For research use only.


Formula: C₁₂H₇NO₃
MDL Number: MFCD00065062
CAS Number: 7797-81-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty