Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-methoxy-4-methylbenzoate 98%

Supplier: AMBEED, INC
Warning

A731117-100G A731117-1G A731117-25G A731117-500G A731117-5G
76727-970EA 66 USD InStock
76727-970 76727-972 76727-974 76727-976 76727-978
Methyl-3-methoxy-4-methylbenzoate 98%
Methyl 3-methoxy-4-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
MDL Number: MFCD00082710
CAS Number: 3556-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty