Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Biphenyl-3-carboxaldehyde 98%

Supplier: AMBEED, INC
Warning

A733638-100MG A733638-1G A733638-5G A733638-250MG
76727-992EA 7 USD InStock
76727-992 76727-994 76727-996 77669-974
Biphenyl-3-carboxaldehyde 98%
Biphenyl-3-carboxaldehyde
Precaución: for Research Use Only


Formula: C₁₃H₁₀O
MW: 182.22 g/mol
MDL Number: MFCD01740432
CAS Number: 1204-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty