Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phorbol 97%

Supplier: AMBEED, INC
Warning

A738144-10MG A738144-1MG A738144-25MG A738144-5MG
76728-034EA 117 USD InStock
76728-034 76728-036 76728-038 76728-040
Phorbol 97%
Phorbol
Precaución: for Research Use Only


Formula: C₂₀H₂₈O₆
MDL Number: MFCD00065450
CAS Number: 17673-25-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty