Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-β-D-glucopyranoside 98%

Supplier: AMBEED, INC
Warning

A738773-1G A738773-25G A738773-5G
76728-048EA 25 USD InStock
76728-048 76728-050 76728-052
Methyl-β-D-glucopyranoside 98%
Methyl-β-D-glucopyranoside
Precaución: for Research Use Only


Formula: C₇H₁₄O₆
MDL Number: MFCD00006602
CAS Number: 709-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty