Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pseudolaric acid B 98%

Supplier: AMBEED, INC
Danger

A739296-100MG A739296-250MG
76728-060EA 168 USD InStock
76728-060 76728-062
Pseudolaric acid B 98%
Pseudolaric acid B
Precaución: for Research Use Only


Formula: C₂₃H₂₈O₈
MDL Number: MFCD01746346
CAS Number: 82508-31-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty