Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-methyl-4-nitropentanoate 98%

Supplier: AMBEED, INC
Warning

A743975-100MG A743975-1G A743975-250MG A743975-5G
76728-094EA 18 USD InStock
76728-094 76728-096 76728-098 77670-010
Methyl-4-methyl-4-nitropentanoate 98%
Methyl-4-methyl-4-nitropentanoate
Precaución: for Research Use Only


Formula: C₇H₁₃NO₄
MDL Number: MFCD00047757
CAS Number: 16507-02-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty