Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Allylbenzylamine 98%

Supplier: AMBEED, INC
Danger

A750534-1G A750534-25G A750534-5G
76728-124EA 17 USD InStock
76728-124 76728-126 77569-420
N-Allylbenzylamine 98%
N-Allylbenzylamine
Precaución: For research use only.


Formula: C₁₀H₁₃N
MDL Number: MFCD00463446
CAS Number: 4383-22-6
UN: 3267
ADR: 8,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty