Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium ((cyclohexylamino)methyl)trifluoroborate 98%

Supplier: AMBEED, INC
Warning

A757457-1G A757457-250MG A757457-5G
76728-174EA 217 USD InStock
76728-174 76728-176 76728-178
Potassium ((cyclohexylamino)methyl)trifluoroborate 98%
Potassium ((cyclohexylamino)methyl)trifluoroborate
Precaución: for Research Use Only


Formula: C₇H₁₄BF₃KN
MDL Number: MFCD10566511
CAS Number: 888711-52-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty