Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Tolyl sulfoxide 98%

Supplier: AMBEED, INC
Warning

A760993-25G A760993-5G A760993-100G
76728-204EA 83 USD InStock
76728-204 76728-206 77670-056
p-Tolyl sulfoxide 98%
p-Tolyl sulfoxide
Precaución: for Research Use Only


Formula: C₁₄H₁₄OS
MDL Number: MFCD00008546
CAS Number: 1774-35-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty