Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-oxotetradecanoate 95%

Supplier: AMBEED, INC
Warning

A765511-1G A765511-250MG A765511-5G
76728-212EA 43 USD InStock
76728-212 76728-214 76728-216
Methyl 3-oxotetradecanoate 95%
Methyl 3-oxotetradecanoate
Precaución: for Research Use Only


Formula: C₁₅H₂₈O₃
MDL Number: MFCD00673474
CAS Number: 22348-97-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty