Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-2-(benzyloxy)phenyltrifluoroborate 95%

Supplier: AMBEED, INC
Warning

A770494-1G A770494-250MG A770494-5G
76728-246EA 55 USD InStock
76728-246 76728-248 76728-250
Potassium-2-(benzyloxy)phenyltrifluoroborate 95%
Potassium 2-(benzyloxy)phenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₁₃H₁₁BF₃KO
MW: 290.13 g/mol
MDL Number: MFCD04115742
CAS Number: 850623-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty