Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-4'-nitrobiphenyl 98%

Supplier: AMBEED, INC
Warning

A789217-1G A789217-25G A789217-5G
76728-420EA 27 USD InStock
76728-420 76728-422 76728-424
4-Hydroxy-4'-nitrobiphenyl 98%
4-Hydroxy-4'-nitrobiphenyl
Precaución: for Research Use Only


Formula: C₁₂H₉NO₃
MW: 215.21 g/mol
MDL Number: MFCD00047742
CAS Number: 3916-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty