Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibutylurea 98%

Supplier: AMBEED, INC
Warning

A801664-25G A801664-5G
76728-572EA 75 USD InStock
76728-572 76728-574
1,3-Dibutylurea 98%
1,3-Dibutylurea
Precaución: for Research Use Only


Formula: H₃CCH₂CH₂CH₂NHC(O)NHCH₂CH₂CH₂CH₃
MDL Number: MFCD00015229
CAS Number: 1792-17-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty