Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phthalamic acid 98%

Supplier: AMBEED, INC
Warning

A814692-100G A814692-10G A814692-1G A814692-25G
76728-668EA 139 USD InStock
76728-668 76728-670 76728-672 76728-674
Phthalamic acid 98%
Phthalamic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₃
MW: 165.15 g/mol
MDL Number: MFCD00025476
CAS Number: 88-97-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty