Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexyltriethoxysilane 98%

Supplier: AMBEED, INC
Warning

A829833-100G A829833-25G A829833-500G A829833-5G
76728-720EA 61 USD InStock
76728-720 76728-722 76728-724 76728-726
Hexyltriethoxysilane 98%
Hexyltriethoxysilane
Precaución: for Research Use Only


Formula: C₁₂H₂₈O₃Si
MW: 248.44 g/mol
MDL Number: MFCD00087744
CAS Number: 18166-37-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty