Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Naproxen sodium 98%

Supplier: AMBEED, INC
Danger

A839347-10MG A839347-25G A839347-50MG A839347-5G
76728-780EA 5 USD InStock
76728-780 76728-782 76728-784 77670-206
Naproxen sodium 98%
Naproxen sodium
Precaución: for Research Use Only


Formula: C₁₄H₁₃NaO₃
MDL Number: MFCD00058507
CAS Number: 26159-34-2
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty