Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PIPES monosodium salt 98%

Supplier: AMBEED, INC
Warning

A844840-1000G A844840-100G A844840-25G A844840-500G
76728-806EA 107 USD InStock
76728-806 76728-808 76728-810 76728-812
PIPES monosodium salt 98%
PIPES monosodium salt
Precaución: for Research Use Only


Formula: C₈H₁₇N₂NaO₆S₂
MDL Number: MFCD00065472
CAS Number: 10010-67-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty