Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Pyridinesulfonic acid 98%

Supplier: AMBEED, INC
Danger

A850998-1G A850998-250MG A850998-5G
76728-854EA 13 USD InStock
76728-854 76728-856 76728-858
2-Pyridinesulfonic acid 98%
2-Pyridinesulphonic acid
Precaución: for Research Use Only


Formula: C₅H₅NO₃S
MW: 159.1631 g/mol
MDL Number: MFCD00234080
CAS Number: 15103-48-7
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty