Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-4-formylphenyltrifluoroborate 98%

Supplier: AMBEED, INC
Warning

A890673-1G A890673-25G A890673-5G
76729-052EA 7 USD InStock
76729-052 76729-054 76729-056
Potassium-4-formylphenyltrifluoroborate 98%
Potassium 4-formylphenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₇H₅BF₃KO
MDL Number: MFCD02093980
CAS Number: 374564-36-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty