Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Octyltrimethylammonium chloride 99%

Supplier: AMBEED, INC
Warning

A897390-100G A897390-25G
76729-076EA 50 USD InStock
76729-076 76729-078
Octyltrimethylammonium chloride 99%
Octyltrimethylammonium chloride
Precaución: for Research Use Only


Formula: C₁₁H₂₆ClN
MW: 207.7838 g/mol
MDL Number: MFCD00059977
CAS Number: 10108-86-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty