Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sarcosine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A897426-100G A897426-25G A897426-500G A897426-5G
76729-080EA 55 USD InStock
76729-080 76729-082 76729-084 76729-086
Sarcosine hydrochloride 95%
Sarcosine hydrochloride
Precaución: for Research Use Only


Formula: C₃H₈ClNO₂
MW: 125.55 g/mol
MDL Number: MFCD00012609
CAS Number: 637-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty