Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triallyl(phenyl)silane 95%

Supplier: AMBEED, INC
Warning

A934215-1G A934215-250MG A934215-5G
76729-246EA 10 USD InStock
76729-246 76729-248 76729-250
Triallyl(phenyl)silane 95%
Triallyl(phenyl)silane
Precaución: for Research Use Only


Formula: C₁₅H₂₀Si
MW: 228.41 g/mol
MDL Number: MFCD00094138
CAS Number: 2633-57-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty