Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl N-(3-methoxy-3-oxopropyl)-N-methyl-beta-alaninate 95%

Supplier: AMBEED, INC
Warning

A937358-1G A937358-250MG A937358-5G
76729-254EA 450 USD InStock
76729-254 76729-256 76729-258
Methyl N-(3-methoxy-3-oxopropyl)-N-methyl-beta-alaninate 95%
Methyl N-(3-methoxy-3-oxopropyl)-N-methyl-beta-alaninate
Precaución: for Research Use Only


Formula: C₉H₁₇NO₄
MDL Number: MFCD00466506
CAS Number: 105-71-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty