Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium oleate 97%

Supplier: AMBEED, INC
Warning

A938990-100G A938990-10G A938990-25G A938990-500G
76729-260EA 36.08 USD InStock
76729-260 76729-262 76729-264 76729-266 76729-268
Sodium oleate 97%
Sodium oleate
Precaución: for Research Use Only


Formula: C₁₈H₃₃NaO₂
MW: 304.45 g/mol
MDL Number: MFCD00004438
CAS Number: 143-19-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty