Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester 98%

Supplier: AMBEED, INC
Warning

A966133-100MG A966133-250MG A966133-5G
76729-342EA 14 USD InStock
76729-342 76729-344 76729-346
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester
Precaución: For research use only.


Formula: C₂₂H₂₅NO₅
MDL Number: MFCD01861331
CAS Number: 110797-35-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty