Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-3-bromopropanesulfonate 98%

Supplier: AMBEED, INC
Warning

A980244-25G A980244-500G A980244-5G
76729-390EA 29 USD InStock
76729-390 76729-392 76729-394
Sodium-3-bromopropanesulfonate 98%
Sodium-3-bromopropanesulfonate
Precaución: for Research Use Only


Formula: C₃H₇BrO₃S
MW: 225.03 g/mol
MDL Number: MFCD00024999
CAS Number: 55788-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty