Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Brimonidine 98%

Supplier: AMBEED, INC
Danger

A103094-10MG A103094-1G A103094-50MG A103094-5G A103094-5MG A103094-100MG A103094-1MG A103094-250MG
76765-272EA 9 USD InStock
76765-272 76765-274 76765-276 76765-278 76765-280 77623-842 77623-844 77623-846
Brimonidine 98%
Brimonidine
Precaución: for Research Use Only


Formula: C₁₁H₁₀BrN₅
MDL Number: MFCD00153878
CAS Number: 59803-98-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty