Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-6-(trifluoromethyl)pyridine-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A109375-10G A109375-1G A109375-250MG A109375-25G A109375-5G
76765-690EA 71 USD InStock
76765-690 76765-692 76765-694 76765-696 76765-698
2-Hydroxy-6-(trifluoromethyl)pyridine-3-carboxylic acid 98%
2-Hydroxy-6-(trifluoromethyl)pyridine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
MDL Number: MFCD01862657
CAS Number: 191595-63-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty