Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-((2-Nitro-1,4-phenylene)bis(azanediyl))diethanol 97%

Supplier: AMBEED, INC
Warning

A110277-10G A110277-1G A110277-250MG A110277-25G
76765-764EA 25 USD InStock
76765-764 76765-766 76765-768 76765-770
2,2'-((2-Nitro-1,4-phenylene)bis(azanediyl))diethanol 97%
2,2'-((2-Nitro-1,4-phenylene)bis(azanediyl))diethanol
Precaución: for Research Use Only


Formula: C₁₀H₁₅N₃O₄
MDL Number: MFCD00239474
CAS Number: 84041-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty