Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,4-Diaminopteridin-6-yl)methanol 95%

Supplier: AMBEED, INC
Warning

A110596-100G A110596-1G A110596-25G A110596-5G
76765-772EA 359 USD InStock
76765-772 76765-774 76765-776 76765-778
(2,4-Diaminopteridin-6-yl)methanol 95%
(2,4-Diaminopteridin-6-yl)methanol
Precaución: for Research Use Only


Formula: C₇H₈N₆O
MDL Number: MFCD00006711
CAS Number: 945-24-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty