Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-N-methyl nicotinamide 96%

Supplier: AMBEED, INC
Warning

A111169-10G A111169-1G A111169-250MG A111169-5G
76765-814EA 79 USD InStock
76765-814 76765-816 76765-818 76765-820
6-Chloro-N-methyl nicotinamide 96%
6-Chloro-N-methyl nicotinamide
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O
MDL Number: MFCD08460991
CAS Number: 54189-82-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty