Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-m-toluonitrile 95%

Supplier: AMBEED, INC
Warning

A111538-100G A111538-10G A111538-1G A111538-25G A111538-5G
76765-846EA 279 USD InStock
76765-846 76765-848 76765-850 76765-852 76765-854
2-Fluoro-m-toluonitrile 95%
2-Fluoro-m-toluonitrile
Precaución: for Research Use Only


Formula: C₈H₆FN
MW: 135.1384 g/mol
MDL Number: MFCD07782078
CAS Number: 185147-07-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty